missing translation for 'onlineSavingsMsg'
Learn More

Ceric Sulfate, 0.1N, Solution, Spectrum™ Chemical

Catalog No. 18609615
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609615 4 L Amber Glass Bottle
18609614 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18609615 Supplier Spectrum Chemical Mfg Cor Supplier No. C1624LTGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Ceric Sulfate Solution
CAS 10294-42-5, 7732-18-5, 7664-93-9
Physical Form Liquid
Quantity 4 L
Grade Ungraded
Concentration or Composition (by Analyte or Components) 0.1 N ± 0.005 N
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.