accessibility menu, dialog, popup

UserName

CH3NNH3COOCH3

CAS RN 10250-61-0

IUPAC Name: methyl 1,5-dimethyl-1H-pyrazole-3-carboxylate
Synonyms: methyl 1,5-dimethylpyrazole-3-carboxylate
Molecular Weight (g/mol): 154.17
Molecular Formula: C7H10N2O2
InChi Key: WOBRRDCECRRRGS-UHFFFAOYSA-N
SMILES: COC(=O)C1=NN(C)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results