accessibility menu, dialog, popup

UserName

OH(R)O(S)ONOO(R)OH(S)OH

CAS RN 10238-27-4

IUPAC Name: (2S,3R,4S,5R)-2-(2-nitrophenoxy)oxane-3,4,5-triol
Synonyms: (2S,3R,4S,5R)-2-(2-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol): 271.23
Molecular Formula: C11H13NO7
InChi Key: YPQCLGUTGDQYNI-DQDDRIPDSA-N
SMILES: O[C@@H]1CO[C@@H](OC2=CC=CC=C2[N+]([O-])=O)[C@H](O)[C@H]1O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)