accessibility menu, dialog, popup

UserName

OCCoCoCOCOCOCOCOCOCO

CAS RN 10210-68-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 341.95
Molecular Formula: C8Co2O8
InChi Key: ZUDHYLQHRXCZGU-UHFFFAOYSA-N
SMILES: [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O=[C-]1[Co][C-](=O)[Co++]1
Physical Form 
  • (3)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)