accessibility menu, dialog, popup

UserName

CAS RN 1020963-78-3

IUPAC Name: N-[(4-fluorophenyl)methyl]-2-methylaniline
Synonyms: N-[(4-fluorophenyl)methyl]-2-methylaniline
Molecular Weight (g/mol): 215.27
Molecular Formula: C14H14FN
InChi Key: JEDDLDDCZKZPOX-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NCC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results