missing translation for 'onlineSavingsMsg'
Learn More

4-(Iodomethyl)tetrahydro-2h-pyran, 97%, Thermo Scientific™

Catalog No. CC29940DE
Change view
Click to view available options
Amber Glass Bottle; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC29940DE Amber Glass Bottle; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC29940DE Supplier Thermo Scientific Chemicals Supplier No. CC29940DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 101691-94-5
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H11IO
Quantity 5g
Synonym 4-iodomethyl tetrahydro-2h-pyran, 4-iodomethyl oxane, 4-iodomethyl-tetrahydro-pyran, 4-iodomethyl tetrahydropyran, 2h-pyran, tetrahydro-4-iodomethyl, 4-iodomethyl-tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-iodomethyl, acmc-20a1ce, 4-iodomethyltetrahydropyran, 4-iodomethyl-tetrahydropyran
InChI Key MQLFSPBSNWUXSO-UHFFFAOYSA-N
SMILES C1COCCC1CI
IUPAC Name 4-(iodomethyl)oxane
Molecular Weight (g/mol) 226.057
PubChem CID 2795507
Formula Weight 226.05
Percent Purity 97%
Chemical Name or Material 4-(Iodomethyl)tetrahydro-2H-pyran
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.