accessibility menu, dialog, popup

UserName

GdONOOONOOONOO

CAS RN 10168-81-7

IUPAC Name: gadolinium(3+) trinitrate
Synonyms: gadolinium(III) nitrate
Molecular Weight (g/mol): 343.26
Molecular Formula: GdN3O9
InChi Key: MWFSXYMZCVAQCC-UHFFFAOYSA-N
SMILES: [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results