accessibility menu, dialog, popup

UserName

CAS RN 10135-38-3

IUPAC Name: 2,2,4,4-tetramethyl-3,4-dihydro-2H-pyrrol-1-ium-1-olate
Synonyms: 2,2,4,4-tetramethyl-3H-pyrrol-1-ium-1-olate
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: GUQARRULARNYQZ-UHFFFAOYSA-N
SMILES: CC1(C)CC(C)(C)[N+]([O-])=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results