accessibility menu, dialog, popup

UserName

CAS RN 1010-26-0

IUPAC Name: 3-oxaspiro[5.5]undecane-2,4-dione
Synonyms: 3-oxaspiro[5.5]undecane-2,4-dione
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: XNDSIASQMRYFSW-UHFFFAOYSA-N
SMILES: O=C1CC2(CCCCC2)CC(=O)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results