accessibility menu, dialog, popup

UserName

CAS RN 101-32-6

HONOH

CAS RN 101-32-6

IUPAC Name: 2-[benzyl(2-hydroxyethyl)amino]ethan-1-ol
Synonyms: ethanol, 2,2'-(benzylimino)di-
Molecular Weight (g/mol): 195.26
Molecular Formula: C11H17NO2
InChi Key: MIZIOHLLYXVEHJ-UHFFFAOYSA-N
SMILES: OCCN(CCO)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results