missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin Related Compound B, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023019
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023019 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023019 Supplier Merck Emd Millipore Supplier No. PHR2229100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 100986-89-8
Melting Point >300°C (literature)
Molecular Formula C13H9F2NO4
Linear Formula C13H9F2NO4
MDL Number MFCD04039905
Quantity 100 mg
Synonym -(-)-9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7 H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
Optical Rotation [α] 20°C/D to 64°C = 1% in DMSO
Formula Weight 281.21
Grade Certified Reference Material