accessibility menu, dialog, popup

UserName

CH3ONHFBr

CAS RN 1009-22-9

IUPAC Name: N-(2-bromo-4-fluorophenyl)acetamide
Synonyms: N-(2-bromo-4-fluorophenyl)acetamide
Molecular Weight (g/mol): 232.05
Molecular Formula: C8H7BrFNO
InChi Key: JAVSBNOXENOHEI-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(F)C=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results