accessibility menu, dialog, popup

UserName

CAS RN 1008402-79-6

ONOOOOOOOOOOOOOONOO

CAS RN 1008402-79-6

IUPAC Name: bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13,16,19,22,25,28-nonaoxahentriacontanedioate
Synonyms: bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13,16,19,22,25,28-nonaoxahentriacontanedioate
Molecular Weight (g/mol): 708.71
Molecular Formula: C30H48N2O17
InChi Key: CQWLSZJYTZVHMU-UHFFFAOYSA-N
SMILES: O=C(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results