accessibility menu, dialog, popup

UserName

CAS RN 100-60-7

IUPAC Name: N-methylcyclohexanamine
Synonyms: methylcyclohexylamine 1-methylcyclohexylamine cyclohexylmethylamine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: XTUVJUMINZSXGF-UHFFFAOYSA-N
SMILES: CNC1CCCCC1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)