missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Extra Thick Glass Fiber (GF) Pre-filters
GREENER_CHOICE

Glass fiber prefilters, depth pre filters, for larger totally filterable volumes in clarification and sterile filtration.

$166.92 - $284.15

Specifications

Autoclavable Autoclavable
Color White
Filter Type Net Filter
Format Disc
For Use With (Equipment) Filtration Systems
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Flow Rate Grade Price Quantity  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Flow Rate Grade Price Quantity  
14-555-451
View Documents Promotion Details
Sartorius
13430130K
5.1 in. 13 cm 348 to 490mL/min. 13430
Pack of 50 Each for $166.92
Only null left
 
14-555-449
View Documents Promotion Details
Sartorius
13430124K
4.88 in. 12.4 cm 348 to 490mL/min. 13430
Pack of 50 Each for $209.63
Only null left
 
14-555-450
View Documents Promotion Details
Sartorius
13430127K
5 in. 12.7 cm 348 to 490mL/min. 13430
Pack of 50 Each for $209.63
Only null left
 
14-555-453
View Documents Promotion Details
Sartorius
1344050Q
1.96 in. 5 cm 4.1 to 4.4 sec. 13440
Pack of 500 Each for $284.15
Only null left
 
Description

Description

The major use of all three glass fibre filters is as a depth prefilter, placed directly on top of a membrane filter, whereby the prefilter diameter specified for the holder must be chosen. Larger diameters would intrude under the sealing ring of the holder and cause leakage. The standard type 13400 contains an acrlic binder. It has a high particle loading capacity, but for very dirty liquids the thicker type 13430 can be more effective. 13440 ia a finer , binder-free type and is recommended for the prefiltration of relative clean solutions such as tissue culture media.Serial filtration may be necessary for difficult-to-filter liquids such as serum.

Specifications

Specifications

Autoclavable
Net Filter
Filtration Systems
Non-sterile
Hydrophilic
500°C
White
Disc
Borosilicate Glass Microfiber
Prefiltration of clean solutions such as tissue culture media
932°F
Glass Microfiber Filter
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'