Learn More
Promega Proteasome-Glo™ Bioluminescent Enzyme Assays
The Proteasome-Glo Assays are homogeneous, luminescent assays that individually measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome in a purified enzyme-based format (not suitable for cell-based applications).
Supplier: Promega G8621
Description
- Measure Chymotrypsin-, Trypsin- and Caspase-Like Activities
- Homogeneous, luminescent enzyme-based assays
- Simple add-mix-measure protocol
- Results in 10-30 minutes after reagent addition
Specifications
| Chymotrypsin-like Assay | |
| Bioluminescence | |
| -30°C to -10°C | |
| 10 mL |
| 1 x 10mL Proteasome Glo Chymotrypsin, 1 x 50μL Suc LLVY Glo substrate, 1 x 1 each Luciferin detection Reagent | |
| 7.2 | |
| Proteasome-Glo | |
| Liquid |