Learn More
Multi-Element Organometallic Oil Standards 23 Element, 300μg/g; SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier:  SPEX CertiPrep Group LLC S23300Y
 View more versions of this product
Catalog No. S23300Y
Only null left
Explore available promotions
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Ag300μg/g
Al300μg/g
B300μg/g
Ba300μg/g
Ca300μg/g
Cd300μg/g
Cr300μg/g
Cu300μg/g
Fe300μg/g
K300μg/g
Mg300μg/g
Mn300μg/g
Mo300μg/g
Na300μg/g
Ni300μg/g
P300μg/g
Pb300μg/g
Sb300μg/g
Si300μg/g
Sn300μg/g
Ti300μg/g
V300μg/g
Zn300μg/g
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
23 Element Oil Standard
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Al, Ba, B
Petrochemical Testing
Triple-leached LDPE bottle
Al,Ba,B
Certified Reference Material
Base Oil 75
100 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Product intended for laboratory use only