Learn More
Gold(III) chloride, 99%
CAS: 13453-07-1 | AuCl3 | 303.32 g/mol
$193.85 - $1806.26
Chemical Identifiers
| CAS | 13453-07-1 |
|---|---|
| Molecular Formula | AuCl3 |
| Molecular Weight (g/mol) | 303.32 |
| MDL Number | MFCD00014172 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC318645000
|
Thermo Scientific Chemicals
318645000 |
500 mg | Glass bottle |
Each for $193.85
|
|
||||
|
AC318640050
|
Thermo Scientific Chemicals
318640050 |
5 g | Glass bottle |
Each for $1,806.26
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13453-07-1 | |
| AuCl3 | |
| 303.32 | |
| MFCD00014172 |
Specifications
| 254.0°C | |
| 13453-07-1 | |
| 500 mg | |
| AuCl3 | |
| MFCD00014172 | |
| Solubility in water: soluble | |
| 303.32 | |
| Glass bottle |
| Brown to Yellow | |
| Crystalline Powder, Crystals and/or Chunks | |
| 98.5% min. (ex Au) | |
| AuCl3 | |
| 15, 4556 | |
| 303.32 | |
| 99% | |
| Gold(III) chloride, 99% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 236-623-1
RTECSNumber : MD5420000
TSCA : TSCA
RUO – Research Use Only