missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride, 99.99+%, (trace metal basis)

CAS: 13453-07-1 | AuCl3 | 303.32 g/mol

$389.11 - $1668.32

Chemical Identifiers

CAS 13453-07-1
Molecular Formula AuCl3
Molecular Weight (g/mol) 303.32
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC437080010
SDS View Documents
Thermo Scientific Chemicals
437080010
1 g Glass bottle
Each for $389.11
Only null left
 
AC437080050
SDS View Documents
Thermo Scientific Chemicals
437080050
5 g Glass bottle
Each for $1,668.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

13453-07-1
AuCl3
303.32

Specifications

254.0°C
13453-07-1
1 g
AuCl3
15,4556
303.32
≥99.99%
Glass bottle
Brown to Red
Crystalline Powder and/or Crystals
63.0 to 66.9% (Au)
AuCl3
Solubility in water: soluble
303.32
Trace Metal Basis
Gold(III) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 236-623-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only