missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bone Sialoprotein II Mouse anti-Human, Clone: ID1.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1061
Description
This Anti-Bone Sialoprotein II Antibody, C-terminus, clone ID1.2 is validated for use in ELISA, RIA, western blot, immunocytochemistry, immunohistochemistry (paraffin) for the detection of Bone Sialoprotein II.Specifications
| Bone Sialoprotein II | |
| Monoclonal | |
| Unconjugated | |
| NM_004967.2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Purified |
| ELISA, Immunocytochemistry, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot | |
| ID1.2 | |
| Purified immunoglobulin from culture supernatant. Lyophilized from 50mM TRIS, pH7.4. | |
| IBSP, BNSP, SP-II, BSP-II, BSP | |
| Bone Sialoprotein isolated from human bones. | |
| Primary | |
| −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
For Research Use Only