Learn More
Thermo Scientific™ Acclaim™ Polar Advantage II HPLC Columns
GREENER_CHOICE
Resolve polar and nonpolar compounds in a single run. These columns have high pH stability (pH 1.5–10) in the separation of basic, neutral, and acidic species.
$464.48 - $2675.23
Specifications
Pore Size 120 Å
Surface Area 300 m²/g
Temperature 60°C
USP Type L60
Stationary Phase Polar Embedded
View More Specs
Products 21
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Particle Size Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Particle Size Price Quantity  
DX063197
View Documents
Thermo Scientific™
063197
150 mm 4.6 mm 5 μm
Each for $1,988.24
Only null left
DX063199
View Documents
Thermo Scientific™
063199
250 mm 4.6 mm 5 μm
Each for $2,172.27
Only null left
078001
View Documents
Thermo Scientific™
078001
100 mm 4.6 mm 3 μm
Each for $2,545.41
Only null left
DX077999
View Documents
Thermo Scientific™
077999
50 mm 2.1 mm 3 μm
Each for $1,400.36
Only null left
DX077998
View Documents
Thermo Scientific™
077998
100 mm 2.1 mm 3 μm
Each for $2,130.83
Only null left
DX077997
View Documents
Thermo Scientific™
077997
250 mm 2.1 mm 3 μm
Each for $2,298.99
Only null left
DX074814
View Documents
Thermo Scientific™
074814
250 mm 2.1 mm 2.2 μm
Each for $2,675.23
Only null left
DX071607
View Documents
Thermo Scientific™
071607
100 mm 3 mm 2.2 μm
Each for $2,275.62
Only null left
03050816
View Documents
Thermo Scientific™
071402
30 mm 2.1 mm 2.2 μm
Each for $1,994.30
Only null left
DX071401
View Documents
Thermo Scientific™
CMD071401
150 mm 2.1 mm 2.2 μm
Each for $2,246.85
Only null left
DX068990
View Documents
Thermo Scientific™
068990
100 mm 2.1 mm 2.2 μm
Each for $2,364.27
Only null left
DX068989
View Documents
Thermo Scientific™
068989
50 mm 2.1 mm 2.2 μm
Each for $1,818.91
Only null left
DX071985
View Documents
Thermo Scientific™
071985
10 mm 3 mm 5 μm
Pack of 2 Each for $698.77
Only null left
DX069699
View Documents
Thermo Scientific™
069699
10 mm 4.6 mm 5 μm
Each for $464.48
Only null left
DX069692
missing translation for 'viewDocuments'
Thermo Scientific™
069692
10 mm 2.1 mm 5 μm
Pack of 2 Each for $827.95
Only null left
DX070080
missing translation for 'viewDocuments'
Thermo Scientific™
070080
250 mm 3 mm 3 μm
Each for $2,581.82
Only null left
DX066276
missing translation for 'viewDocuments'
Thermo Scientific™
066276
33 mm 3 mm 3 μm
Each for $1,338.91
Only null left
DX063705
missing translation for 'viewDocuments'
Thermo Scientific™
063705
150 mm 3 mm 3 μm
Each for $2,240.08
Only null left
DX063191
missing translation for 'viewDocuments'
Thermo Scientific™
063191
150 mm 4.6 mm 3 μm
Each for $1,951.22
Only null left
DX063189
missing translation for 'viewDocuments'
Thermo Scientific™
063189
50 mm 4.6 mm 3 μm
Each for $1,299.98
Only null left
DX063187
missing translation for 'viewDocuments'
Thermo Scientific™
063187
150 mm 2.1 mm 3 μm
Each for $1,988.24
Only null left
Description
Description
Resolve polar and nonpolar compounds in a single run with Thermo Scientific™ Acclaim™ PolarAdvantage II (PA2) reversed-phase columns. These high-efficiency, silica-based columns have a polar-embedded stationary phase that operates over a wider range of chromatographic conditions than possible with conventional reversed-phase stationary phases. Its unique chemistry provides enhanced hydrolytic stability from pH 1.5 to 10 with 100% aqueous mobile phases. The column exhibits selectivity that is complementary to conventional C18 columns and excellent peak shapes for both basic and acidic compounds.
Complementary Selectivity and Enhanced Hydrolytic Stability
  • Compatible with 100% aqueous mobile phases
  • High reversed-phase capacity
  • Exceptional hydrolytic stability from pH 1.5 to 10
  • Extremely low bleed, fully compatible with mass spectrometry (MS)
  • Available in 2.2, 3, and 5μm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 120Å
  • Also available in 2.2μm RSLC format for reliable, fast separations
About the Acclaim Family of HPLC Columns
Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These ultrapure silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
  • Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations
Specifications
Specifications
120 Å
60°C
Polar Embedded
Reversed Phase
Reversed Phase
300 m²/g
L60
Spherical, Fully Porous, Ultrapure Silica
Yes
Videos
SDS
Documents
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.