Learn More
Research Products Zymoprep Yeast Plasmid Miniprep II w/Zymo-Spin I Columns, Uncapped, 50 Preps
Supplier: Research Products Corporation ZD2004
The Zymoprep Yeast Plasmid Miniprep II is a simple and efficient yeast plasmid miniprep that is based on the E. coli alkaline lysis method but using Zymolyase as the first solution. The kit provides all the necessary reagents for plasmid isolation from S. cerevisiae, C. albicans and S. pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The procedure is simple and efficient, with no need for glass beads or phenol. The Zymoprep II recovers about 5 fold more plasmid DNA compared to Zymoprep I and eliminates the isopropanol precipitation step. Reliably recover plasmid DNA from yeast colonies, patches on plates, or as liquid cultures. The system is ideal for low-copy number and hard-to-isolate plasmids. Eluted plasmid DNA can be used directly for E. Coli transformation, PCR, and Southern blot analysis.
Simple: Quickly and easily rescue plasmid from yeast.