Learn More
Research Products Zymo-Spin IIC Columns, Capped, 250/pack DFS Item
Supplier:  Research Products Corporation ZC1011250
The Zymo-Spin IIC Fast-Spin column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin IIC features durable polypropylene construction with an attached cap and contains a unique silica-based matrix that allows purification of up to 25 g of DNA or 50 g of RNA, in 25 l eluate. Capacity is 900 l.
Catalog No. 501977364
Only null left
Explore available promotions