Learn More
Research Products Zymo-Spin I Columns, Uncapped, 250/pk
Supplier: Research Products Corporation ZC1003250
The Zymo-Spin I Fast-Spin column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin I features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g DNA or RNA in 6 l eluate. Capacity is 800 l.