missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 200 μL

Catalog No. 14222924
Change view
Click to view available options
Tip Style:
Standard, Round
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
14222924 Standard, Round
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222924 Supplier Axygen™ Supplier No. ZTF200RS

Clear, autoclavable tips; color-coded racks.

  • Clear Tips
  • Format: 96 well
  • Propylene construction
  • Certified RNase/DNase free
  • Nonpyrogenic
  • Aerosol resistant filters
  • Sterile
  • Compatibility: Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

Specifications

Volume (Metric) 200 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
Format Rack
Tip Style Standard, Round
Packaging Non-SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Product Line Caliper Zymark
Material Polyethylene
Show More Show Less