missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 200 μL
Clear, autoclavable tips; color-coded racks.
Supplier: Axygen™ ZTF200LRS
Description
- Clear Tips
- Format: 96 well
- Propylene construction
- Certified RNase/DNase free
- Nonpyrogenic
- Aerosol resistant filters
- Sterile
- Compatibility: Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head
Specifications
| 200 μL | |
| Sterile | |
| Blue | |
| Non-ANSI | |
| Rack | |
| Non-SBS Rack | |
| Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head | |
| Polyethylene |
| Filtered | |
| Clear | |
| Autoclavable | |
| Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss | |
| Maxymum Recovery, Round | |
| 50 Racks/Cs. | |
| Caliper Zymark |