missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ZWITTERGENT™ 3-12 Detergent, Calbiochem™,

Catalog No. 6930155GM
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
Plastic Bags
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6930155GM 5 g Plastic Bags
69301525GM 25 g Glass Bottle
6930151KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6930155GM Supplier MilliporeSigma™ Supplier No. 6930155GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14933-08-5
Molecular Formula C17H37NO3S
Molecular Weight (g/mol) 335.547
InChI Key IZWSFJTYBVKZNK-UHFFFAOYSA-N
Synonym n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
PubChem CID 84703
ChEBI CHEBI:75303
IUPAC Name 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Color White
Quantity 5 g
Packaging Plastic Bags
Percent Purity ≥99% (TLC)