missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ZWITTERGENT™ 3-12 Detergent, Calbiochem™,

Supplier:  MilliporeSigma™ 69301525GM

Quantity 25 g
Packaging Glass Bottle

 View more versions of this product

Catalog No. 69301525GM


Only null left

Description

Description

Specifications

Chemical Identifiers

14933-08-5
335.547
n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
CHEBI:75303
CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
C17H37NO3S
IZWSFJTYBVKZNK-UHFFFAOYSA-N
84703
3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate

Specifications

White
25 g
C17H37NO3S
IZWSFJTYBVKZNK-UHFFFAOYSA-N
3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate
84703
≥99% (TLC)
14933-08-5
Glass Bottle
n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
335.547
CHEBI:75303
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions