missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ IPGRunner™ Mini-cell

Catalog No. ZM0001
Encompass
Change view
Click to view available options
Quantity:
1 system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ZM0001 1 system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZM0001 Supplier Invitrogen™ Supplier No. ZM0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

ZOOM IPGRunner Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini-Cell chamber

Oil-free, trouble-free 2D electrophoresis in a minigel format

The ZOOM IPGRunner System consists of three main components. The ZOOM IPGRunner Mini-Cell includes a high-voltage electrophoresis core and lid assembly that fits a mini-cell chamber identical to the XCell SureLock™ mini-cell. The mini-cell holds two ZOOM IPGRunner Cassettes. The ZOOM IPGRunner Cassettes (available separately) are provided ready to use and self-contained, eliminating oil overlays for convenient sample rehydration and IEF. Each cassette holds up to six ZOOM Strips. The ZOOM Strips (available separately) are 7cm long and contain a thin layer of polyacrylamide gel that includes a fixed pH gradient. Each ZOOM Strip is clearly labeled with a unique identifying number, pH range, and orientation marks. ZOOM Strips are supplied attached to a tri-fold card for easy access and removal.

  • The ZOOM IPGRunner System typically completes first dimension separation isoelectric focusing (IEF) in less than three hours
  • The minigel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage

Quick and Simple Set-up Procedure

  • Easily load samples and set up the ZOOM IPGRunner System for IEF
  • Once IEF is complete, the ZOOM IPGRunner Mini-Cell can be used with NuPAGE™ and Novex™ ZOOM™ gels to complete SDS-PAGE analysis in 40 minutes

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Specifications

Content And Storage The ZOOM IPGRunner Mini-Cell includes the ZOOM IPGRunner Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber.
For Use With (Equipment) XCell SureLock™ Mini, ZOOM™ IPGRunner™ Mini
Gel Compatibility ZOOM™ IPG Strips
Gel Size 7 cm Strips
Product Line IPGRunner, ZOOM
Quantity 1 system
What is the protocol for using ZOOM gels after first-dimension separation on an IPG strip?

1) Trim excess plastic from IPG strip

2) Equilibrate IPG strip for 2 x 15 min in 5 mL of the appropriate buffer (Use NuPAGE sample buffer for NuPAGE ZOOM gels and Tris Glycine ZOOM gels). Equilibration buffer: 1X NuPAGE sample buffer, 50 mM DTT.

3) Place equilibrated IPG strip in large well of ZOOM gel. Seal the strip in place with an agarose (0.5%) overlay.

4) Load molecular weight markers in small well of ZOOM gel.

Variations: To alkylate the proteins after reducing them, prior to separation on ZOOM gels: 1) Incubate IPG strip for 15 min in equilibration buffer 2) Transfer strip to Equilibration Buffer containing 125 mM iodoacetamide and lacking reducing agent. Incubate an additional 15 min. To denature the proteins with urea after IEF: 6 M urea can be added to Equilibration Buffer, if desired.

After the first dimension separation using the ZOOM IPGRunner, I do not see any distinct spots. What could the issue be?

Here are the possible causes and solutions:

- Low protein load. Increase the protein load. Use an accurate and sensitive protein estimation method.
- Improper sample preparation. Increase solubilization reagents. Use at least 8 M urea for solubilization. Add DTT and non-ionic detergents (see manual for details)
- Strip not correctly oriented. Align the strip correctly as described in the manual. Be sure to have the gel side up when loading the strip into the ZOOM IPGRunner Cassette.
- Air bubbles between the strip and 2D gel. Smooth out any air bubbles.
- Insensitive detection method. Use sensitive detection methods such as silver staining or immunoblotting.

I just set up the ZOOM IPGRunner for IEF but don't see any current running through the system. What is the issue?

A possible reason is poor contact between electrodes or incomplete circuit. Make sure that you have added 600 µL deionized water to the Electrode Wicks and the gel is exposed at the anodic and cathodic ends of the cassette. Check the power supply. Be sure to set the “Load Check” to off to enable the power supply to operate at low current.

What are the power supply requirements for the ZOOM IPGRunner?

The ZOOM IPGRunner System should be used with an external DC power supply designed for electrophoresis applications. This power supply must:

- Be isolated from the ground so that the DC output is floating (safety requirement).
- Be programmable, with a 4-protocol minimum.
- Be able to operate at low current (less than 1 mA) as IEF is performed at very low current
Note: Many power supplies automatically shut off when the current drops below 1 mA. You will need a power supply capable of overriding the low current shut-off feature. The electrical leads of the ZOOM IPGRunner Lid are recessed and may not fit into some power supply units. To allow connection of the ZOOM IPGRunner power leads with certain power supplies, use Invitrogen Power Supply Adapters available separately (Cat. No. ZA10001).

What is the tolerance of the ZOOM IPGRunner Mini-Cell to organic solvents?

The ZOOM IPGRunner Mini-Cell is impervious to alcohol, but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene) or acetone.

What do you recommend for regular maintenance of the ZOOM IPGRunner Mini-Cell?

We recommend washing the ZOOM IPGRunner Mini-Cell with a mild detergent and rinsing with deionized water after each use. Do not wash lid with cables plugged into the power supply.

How can I adapt my XCell SureLock Mini-Cell for two-dimensional (2D) electrophoresis using your ZOOM IPGRunner system?

We offer the ZOOM IPGRunner Retrofit kit for adapting the XCell SureLock Mini-Cell for two-dimensional (2D) electrophoresis.

What are the components of the ZOOM IPGRunner Mini-Cell?

Here are the components of the ZOOM IPGRunner Mini-Cell:

ZOOM IPGRunner Core
ZOOM IPGRunner Lid
Gel Tension Wedge
Buffer Dam
Mini-Cell Chamber

I used the ZOOM IPGRunner system and some of my protein spots are missing or appear as a smear. Can you please offer some suggestions?

Here are the possible causes and solutions:

*Protein degradation: Add protease inhibitors during sample preparation (see manual for details).
*Different subunits: Use denaturing conditions (8 M urea).
*Incomplete equilibration: Perform equilibration as described in the manual. Increase the equilibration time.
*Protein precipitates: Increase solubilization reagents in the rehydration buffer (see manual for details. Use appropriate strips based on the pI of the protein sample.
*Low protein load: Increase the protein load. You can load up to 400 µg of fractionated protein sample per ZOOM Strip. Use an accurate and sensitive protein estimation method.
*Insensitive detection method: Use sensitive detection methods such as silver staining or immunoblotting.

I used the ZOOM IPGRunner system and got vertical streaking. Can you please offer some suggestions?

Here are the possible causes and solutions:

*Protein has got oxidized: Include DTT in the rehydration buffer and perform the alkylation step.
*Impure solutions: Use ultrapure reagents to prepare the rehydration buffer, equilibration buffer, and buffers for SDS/PAGE.

I used the ZOOM IPGRunner System and got horizontal streaking. Can you please offer some suggestions?

Here are the possible causes and solutions:

*Impure solutions: Use ultrapure reagents to prepare the rehydration buffer, equilibration buffer, and buffers for SDS/PAGE.
*Air bubble between the strip and 2D gel: Smooth out the air bubbles.
*Poor strip rehydration: Rehydrate the strips in 140 µL rehydrating buffer for 1 hour as described in the manual. Rehydration can be extended to overnight if you use 155 µL rehydrating buffer. Make sure that the rehydration buffer covers the strip completely.
*Protein overload: Decrease the protein concentration or lower the sample volume.
*Protein precipitates: Increase solubilization reagents in the rehydration buffer (see manual). Use appropriate strips based on the pI of the protein sample. Do not add more than 10 µL of your sample to 140 µL of rehydration buffer (see manual).

I used the ZOOM IPGRunner System and got incomplete focusing of my proteins. Can you please offer some tips?

Here are the possible causes and solutions:

*Improper sample preparation: Increase solubilization reagents in the rehydration buffer. Use 8 M urea for solubilization. Add DTT and non-ionic detergents as described in the manual.
*Incorrect focusing time: Increase or decrease the focusing time based on the initial results.
*High protein load: Decrease the protein load. Use an accurate and sensitive protein estimation method.
*Salt, lipids, or nucleic acid impurities present in the sample: Remove interfering substances (see manual for details).
*Poor strip rehydration: Rehydrate the strips in 140 µL rehydrating buffer for 1 hour as described in the manual. Rehydration can be extended to overnight if you use 155 µL rehydrating buffer. Make sure the rehydration buffer is covering the strip completely.

After the first dimension separation using the ZOOM IPGRunner System, I do not see any distinct spots. What could the issue be?

Here are the possible causes and solutions:

*Low protein load. Increase the protein load. Use an accurate and sensitive protein estimation method.
*Improper sample preparation. Increase solubilization reagents. Use at least 8 M urea for solubilization. Add DTT and non-ionic detergents (see manual for details).
*Strip not correctly oriented. Align the strip correctly as described in the manual. Be sure to have the gel side up when loading the strip into the ZOOM IPGRunner Cassette.
*Air bubbles between the strip and 2D gel. Smooth out any air bubbles.
*Insensitive detection method. Use sensitive detection methods such as silver staining or immunoblotting.

What are the power supply considerations for the ZOOM IPGRunner system?

The ZOOM IPGRunner System should be used with an external DC power supply designed for IEF and electrophoresis applications. This power supply must:

*Be isolated from the ground so that the DC output is floating.
*Be programmable, with a 4-protocol minimum.
*Be able to operate at low current (<1 mA) as IEF is performed at very low current.
Note: Many power supplies automatically shut off when the current drops below 1 mA. You will need a power supply capable of overriding the low current shut-off feature.

The electrical leads of the ZOOM IPGRunner Lid are recessed and may not fit into some power supply units. To allow connection of the ZOOM IPGRunner power leads with certain power supplies, use Invitrogen Power Supply Adapters available separately (Cat. No. ZA10001).

What is the optimal concentration of detergent that I should add to my sample for the ZOOM IPG Runner?

Detergents are used in the sample as solubilizing agents:

*SDS must be 0.2% or lower. If the percentage of SDS is too high, all the proteins will be negatively charged and will migrate to the positive electrode.
*CHAPS can be increased up to 4%.
*NP-40, Triton-X and other nonionic detergents are used at 0.5-0.7%.

What is the optimal salt concentration that I should use in my sample for the ZOOM IPG Runner?

The salt concentration should not exceed 10 mM for these reasons:

*Burning can occur when conductivity is high, hence current limits should be set.
*Protein transport in an electric field decreases with increasing ionic strength.
*The efficiency of all IEF instruments is decreased when samples contain more than 10 mM salt.
*Longer run times are generally required for samples containing >10 mM salt.

Can I run non-reduced samples in the ZOOM IPGRunner?

You can run non-reduced samples in the ZOOM IPGRunner by omitting DTT in the rehydration buffer and equilibration buffers.

Can I heat my protein sample prepared using ZOOM 2D Protein Solubilizer?

We do not recommend heating protein samples containing urea over 37 degrees C as elevated temperatures cause urea to hydrolyze to isocyanate, which modifies proteins by carbamylation.

How do you recommend storing protein samples prepared in the Sample Rehydration buffer?

We recommend storing them at -80 degrees C. We do not recommend storing them at -20 degrees C

How much ampholyte do you recommend adding to the sample rehydration buffer?

The recommended ampholyte concentration in the sample rehydration buffer is 0.5%.

*If you are loading 5-50 µg of protein (pure protein or crude lysate) per ZOOM strip, use 0.5% ampholytes in the sample rehydration buffer.
*If you are loading >50 µg of protein (crude lysate or fractionated sample) per ZOOM Strip, use 0.5-2% ampholytes in the sample rehydration buffer.

Note: Higher ampholyte concentration requires longer focusing times.

What is the maximum recommended volume of protein sample to be added to the Sample Rehydration buffer?

The maximum volume of the protein sample should at most be 1/6 of the final sample volume that will be added to the strip. A good general volume would be 5-10µL. 140 µL of sample diluted in Sample Rehydration buffer is used to rehydrate each ZOOM Strip for the standard rehydration time of one hour. For overnight rehydration, we recommend using 155 µL of diluted sample.

I am using your ZOOM IPGRunner system. What is the sample rehydration buffer?

The sample rehydration buffer, also known as the sample buffer, is used to denature and solubilize protein samples, and rehydrate the IPG strips prior to performing IEF using the ZOOM IPGRunner system. Due to the large variety of proteins, there is no universal sample rehydration buffer. The sample rehydration buffer must maintain proteins in solution during rehydration of the IPG strips and IEF, and must not have any effect on the pI of the protein. The buffer typically contains a denaturing agent (urea or urea/thiourea), solubilizing agent (non-ionic or zwitterionic detergent and ampholytes), and reducing agent (DTT).

Note: To obtain high resolution, IEF is usually performed under denaturing and reducing conditions. Use a denaturing sample rehydration buffer for sample preparation (see manual for recipe).

How does the ZOOM IPG Runner System work?

The ZOOM IPGRunner System provides a convenient and quick way to perform isoelectric focusing (IEF) of proteins in a vertical mini-gel format using immobilized pH gradient (IPG) strips for two- dimensional (2D) gel electrophoresis.

The major components of the ZOOM IPGRunner System are:

*ZOOM IPGRunner Mini-Cell (Cat. No. ZM0001)
*ZOOM IPGRunner Cassettes (1 pack of 10 cassettes, Cat. No. ZM0003)
*ZOOM Strips (12 ZOOM Strips, pH 3–10 NL, Cat. No. ZM0011)

Using the ZOOM Strips, proteins are separated based on their isoelectric point (pI). The proteins in the sample can be further separated in the second dimension, based on their molecular weight, by placing the IPG Strips into the IPG well of a NuPAGE Bis-Tris ZOOM gel or Tris-Glycine ZOOM gel. The proteins separated in the second dimension can be visualized as spots by the use of SimplyBlue SafeStain or silver staining, or blotted onto membranes. Protein spots can be also excised from the gel or the membranes to be further analyzed by mass spectrometry or chemical microsequencing to facilitate protein identification.

What is the purpose of alkylation prior to IEF?

Alkylation prevents unwanted protein modifications by alkylating cysteines to avoid mixed disulfide formation and reoxidation and this allows for crisper focusing.

Do you still offer ZOOM 2D Protein Solubilizers?

Sorry we have discontinued selling the ZOOM 2D Protein Solubilizers.

What are the main reasons for performing 2D gel electrophoresis of proteins?

The main advantages of performing 2D gel electrophoresis of proteins and applications used for are listed below:

Advantages:

*Simultaneous separation of hundreds to thousands of proteins
*High capacity with superior resolution
*Compatible with further analysis by MS for protein identification and sequencing
Ability to separate and analyze low-abundance proteins

Applications:

*Comparative proteomics: identifying and analyzing differences between complex mixtures of proteins
*Protein profiling, biomarker discovery
*Separation and analysis of protein variants and isoforms

What products do you offer for 2D gel electrophoresis of proteins?

We offer the following products for the first- and second-dimension separation of proteins:

First-dimension separation:

*Vertical gels for separation of proteins based on their isoelectric point (pI) (https://www.thermofisher.com/us/en/home/life-science/protein-expression-and-analysis/protein-gel-electrophoresis/protein-gels/specialized-protein-separation/isoelectric-focusing.html)

*Solution-phase isoelectric focusing of proteins using ZOOM Disks (immobilized buffer disks of specific pH) to reduce sample complexity, enrich low-abundance proteins, and increase the dynamic range of detection (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/isoelectric-focusing/zoom-ief-fractionator.html)

*Mini gel system for high-throughput isoelectric focusing of proteins using ZOOM IPG (Immobilized pH Gradient) Strips (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/2d-gel-electrophoresis/zoom-ipgrunner-system.html)

Second-dimension separation:

*ZOOM gels for 2D electrophoresis: NuPAGE Bis-Tris (Cat. No. NP0330BOX) and Tris-Glycine (Cat. No. EC60261BOX) mini gels with IPG wells ( to accommodate 7 cm ZOOM strips) for separation of proteins based on their molecular weight

What is 2D protein gel electrophoresis?

2D protein gel electrophoresis is the separation of proteins in two dimensions. In the first dimension, proteins are separated by their isoelectric point (pI) using isoelectric focusing, and in the second dimension, they are separated by their mass using SDS-PAGE.


For Research Use Only. Not for use in diagnostic procedures.