missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ IPGRunner™ Combo Kit

Catalog No. ZM0002
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ZM0002 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZM0002 Supplier Invitrogen™ Supplier No. ZM0002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

ZOOM IPGRunner Mini-Cell, electrode wicks, sealing tape, 10 ZOOM IPGRunner Cassettes, and 12 ZOOM Strips, pH 3-10 NL

Oil-free, 2D electrophoresis in a minigel format

The ZOOM IPGRunner System typically completes first dimension separation isoelectric focusing (IEF) in less than three hours. The minigel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage.

  • The ZOOM IPGRunner Combo Kit includes everything needed for first dimension separation: ZOOM IPGRunner Mini-Cell; 10 ZOOM IPGRunner Cassettes; 12 ZOOM Strips, pH 3-10NL
  • For second dimension separation, Novex™ ZOOM gels and the XCell SureLock™ Mini-Cell are recommended (both are available separately)

ZOOM IPGRunner Mini-Cell

  • Consists of a ZOOM IPGRunner Core, a mini-cell chamber, and a lid
  • Can be used to perform isoelectric focusing of up to 12 IPG strips (ZOOM Strips) loaded in two ZOOM IPGRunner Cassettes
ZOOM IPGRunner Cassette
  • Used to perform oil-free rehydration and isoelectric focusing of up to six 7.0cm IPG strips (ZOOM Strips) without requiring separate units for rehydration and isoelectric focusing
  • Disposable
ZOOM Strips
  • 7cm long
  • Contain a thin layer of polyacrylamide gel with a fixed pH gradient
  • Clearly labeled with a unique identifying number, pH range, and orientation marks
  • Supplied attached to a tri-fold card for easy access and removal
Novex ZOOM Gels (available separately)
  • ZOOM Gels are precast polyacrylamide gels designed for 2D analysis of proteins following isoelectric focusing with ZOOM Strips
  • They are recommended for use with our XCell SureLock™ Mini-Cell
  • ZOOM Gels contain an IPG well and a molecular weight marker well; the IPG well is designed to accommodate the 7.0cm ZOOM Strips
  • Two types of ZOOM Gels are available: NuPAGE™ Novex 4–12% Bis-Tris ZOOM Gel and Novex 4–20% Tris-Glycine ZOOM Gel

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Specifications

Content And Storage The ZOOM™ IPGRunner™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini-Cell chamber

The ZOOM™ IPGRunner™ Combo Kit includes the ZOOM™ IPGRunner™ Mini-Cell, electrode wicks, sealing tape, 10 ZOOM™ IPGRunner™ Cassettes, and 12 ZOOM™ Strips, pH 3-10 NL

The ZOOM™ IPGRunner™ Retrofit Kit for XCell SureLock™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid. The ZOOM™ IPGRunner™ Cassettes include electrode wicks and sealing tape
For Use With (Equipment) XCell SureLock Mini, ZOOM IPGRunner Mini
Gel Compatibility ZOOM™ IPG Strips
Gel Size 7 cm Strips
Product Line IPGRunner, ZOOM
Quantity 1 kit
What may cause streaking on the 2nd dimension gel after IEF?

There are several reasons why streaking may occur.

(1) Sample is not completely solubilized prior to application.

(2) Sample is poorly soluble in rehydration solution.

(3) Non-protein impurities in the sample can interfere with IEF, causing horizontal streaking in the final 2-D result, particularly toward the acidic side of the gel.

(4) Ionic impurities are present in sample.

(5) Ionic detergent is present in sample.

(6) Sample load is too high.

(7) Underfocusing. Focusing time was not long enough to achieve steady state focusing.

(8) Overfocusing. Extended focusing times (over 100,000 Vh) may result in electroendosmotic water and protein movement, which can produce horizontal smearing.

What should be done?

(1) Be sure that the sample is completely and stably solubilized. Note: Repeated precipitation-resolubilization cycles produce or increase horizontal streaking.

(2) Increase the concentration of the solubilizing components in the rehydration solution.

(3) Modify sample preparation to limit these contaminants or dialyze protein.

(4) Reduce salt concentration to below 10 mM by dilution or desalt the sample by dialysis. Precipitation with TCA and acetone and subsequent resuspension is another effective desalting technique that removes lipids, nucleotides and other small molecules.
Note: Specific and non-specific losses of proteins can occur with dialysis, gel chromatography, and precipitation/resuspension of samples. If the sample preparation cannot be modified, the effect of ionic impurities can be reduced by modifying the IEF protocol. Limit the voltage to 100-150 V for 2 hours, then resume a normal voltage step program. This pre-step allows the ions in the sample to move to the ends of the IPG strip.

(5) If the ionic detergent SDS is used in sample preparation, the final concentration must not exceed 0.25% after dilution into the rehydration solution. Additionally, the concentration of the non-ionic detergent present must be at least 8 times higher than the concentration of any ionic detergent to ensure complete removal of SDS from the proteins.

(6) Extend focusing time. Load less sample.

(7) Prolong focusing time.

(8) Reduce focusing time.

Will my proteins focus better on the ZOOM IPGRunner system if I use a voltage higher than 2000 V?

With the ZOOM system, it is not recommended to use a voltage higher than 2000 V. The ZOOM IPG Runner System is rated to 3500 VDC and 3.5 Watts, but for performing IEF, the results are optimal at maximum of 2000 V and 0.1 W per strip (~50 µA per strip).

My sample is in Sample Rehydration Buffer containing urea. What would happen if it is heated?

To avoid modification of proteins, never heat a sample over 37 degrees C after adding urea. Elevated temperatures cause urea to hydrolyze to isocyanate, which modifies proteins by carbamylation.

What is the maximum amount of SDS that can be in a sample for the ZOOM IPGRunner System?

We do not recommend using more than 0.2% SDS. If the percentage of SDS is too high, all the proteins will be negatively charged and will migrate to the positive electrode.

What is the recommended amount of protein to load on the IPG strip?

A typical load amount is 5-10 µg for silver staining and 10-50 ug for Coomassie staining. We have determined that 400 µg of protein can be focused on our 7 cm strips with a mixture of purified standard proteins. To load this much protein, some cleaning up (like pre-fractionation) of crude lysate is required. Often this is because of the large dynamic range between "high abundance" proteins vs "low abundance" proteins, making the most abundant proteins the load-limiting factor. This is cell/tissue dependent. When testing sample preparation and run parameters, we recommend that the typical load recommended above for the stain of choice be used. After satisfactory results are obtained, higher loads can be run. Longer run times may be required when using higher loads.

Can I store a sample that I have prepared in the Sample Rehydration Buffer (for the ZOOM IPGRunner System) for later IPG strip rehydration?

You may store a sample made up in the ZOOM 2D Protein Solubilizer or Sample Rehydration Buffer at -80 degrees C. Storage at -20 degrees C is not recommended.

What is the maximum volume of protein sample that can be included with the Sample Rehydration Buffer?

A good general volume would be 5-10 µL, but more protein sample volume can be loaded. The protein sample should be no more than 1/6 of the final volume that will be added to the strip. You will need 140 µL of sample diluted in sample rehydration buffer to rehydrate the ZOOM strips for the standard rehydration time of one hour. If you want to extend the rehydration time to overnight, you will need a total of 155 µL. If the desired amount of protein is too low to be loaded into the strip at this volume and be adequately visualized, the protein sample may need to be concentrated.

Is it possible to stain the ZOOM IPG strip after running the first dimension?

Staining the ZOOM IPG strips is generally not recommended. If your protein sample is a cell lysate, you will get a smear. It may work with purified or simple protein mixtures. You can stain standards if you have strips to spare. A protocol for staining the IPG strips after the first dimension IEF can be found in ZOOM IPGRunner System user guide (search for ZM0001 on the Thermo Fisher Scientific website). At this time, however, it has not been determined if this protocol is compatible with subsequent mass spectrometry.

What is the protocol for using ZOOM gels after first-dimension separation on an IPG strip?

1) Trim excess plastic from IPG strip

2) Equilibrate IPG strip for 2 x 15 min in 5 mL of the appropriate buffer (Use NuPAGE sample buffer for NuPAGE ZOOM gels and Tris Glycine ZOOM gels). Equilibration buffer: 1X NuPAGE sample buffer, 50 mM DTT.

3) Place equilibrated IPG strip in large well of ZOOM gel. Seal the strip in place with an agarose (0.5%) overlay.

4) Load molecular weight markers in small well of ZOOM gel.

Variations: To alkylate the proteins after reducing them, prior to separation on ZOOM gels: 1) Incubate IPG strip for 15 min in equilibration buffer 2) Transfer strip to Equilibration Buffer containing 125 mM iodoacetamide and lacking reducing agent. Incubate an additional 15 min. To denature the proteins with urea after IEF: 6 M urea can be added to Equilibration Buffer, if desired.

After the first dimension separation using the ZOOM IPGRunner, I do not see any distinct spots. What could the issue be?

Here are the possible causes and solutions:

- Low protein load. Increase the protein load. Use an accurate and sensitive protein estimation method.
- Improper sample preparation. Increase solubilization reagents. Use at least 8 M urea for solubilization. Add DTT and non-ionic detergents (see manual for details)
- Strip not correctly oriented. Align the strip correctly as described in the manual. Be sure to have the gel side up when loading the strip into the ZOOM IPGRunner Cassette.
- Air bubbles between the strip and 2D gel. Smooth out any air bubbles.
- Insensitive detection method. Use sensitive detection methods such as silver staining or immunoblotting.

I just set up the ZOOM IPGRunner for IEF but don't see any current running through the system. What is the issue?

A possible reason is poor contact between electrodes or incomplete circuit. Make sure that you have added 600 µL deionized water to the Electrode Wicks and the gel is exposed at the anodic and cathodic ends of the cassette. Check the power supply. Be sure to set the “Load Check” to off to enable the power supply to operate at low current.

What are the power supply requirements for the ZOOM IPGRunner?

The ZOOM IPGRunner System should be used with an external DC power supply designed for electrophoresis applications. This power supply must:

- Be isolated from the ground so that the DC output is floating (safety requirement).
- Be programmable, with a 4-protocol minimum.
- Be able to operate at low current (less than 1 mA) as IEF is performed at very low current
Note: Many power supplies automatically shut off when the current drops below 1 mA. You will need a power supply capable of overriding the low current shut-off feature. The electrical leads of the ZOOM IPGRunner Lid are recessed and may not fit into some power supply units. To allow connection of the ZOOM IPGRunner power leads with certain power supplies, use Invitrogen Power Supply Adapters available separately (Cat. No. ZA10001).

What is the tolerance of the ZOOM IPGRunner Mini-Cell to organic solvents?

The ZOOM IPGRunner Mini-Cell is impervious to alcohol, but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene) or acetone.

What do you recommend for regular maintenance of the ZOOM IPGRunner Mini-Cell?

We recommend washing the ZOOM IPGRunner Mini-Cell with a mild detergent and rinsing with deionized water after each use. Do not wash lid with cables plugged into the power supply.

How can I adapt my XCell SureLock Mini-Cell for two-dimensional (2D) electrophoresis using your ZOOM IPGRunner system?

We offer the ZOOM IPGRunner Retrofit kit for adapting the XCell SureLock Mini-Cell for two-dimensional (2D) electrophoresis.

What are the components of the ZOOM IPGRunner Mini-Cell?

Here are the components of the ZOOM IPGRunner Mini-Cell:

ZOOM IPGRunner Core
ZOOM IPGRunner Lid
Gel Tension Wedge
Buffer Dam
Mini-Cell Chamber

I used the ZOOM IPGRunner system and some of my protein spots are missing or appear as a smear. Can you please offer some suggestions?

Here are the possible causes and solutions:

*Protein degradation: Add protease inhibitors during sample preparation (see manual for details).
*Different subunits: Use denaturing conditions (8 M urea).
*Incomplete equilibration: Perform equilibration as described in the manual. Increase the equilibration time.
*Protein precipitates: Increase solubilization reagents in the rehydration buffer (see manual for details. Use appropriate strips based on the pI of the protein sample.
*Low protein load: Increase the protein load. You can load up to 400 µg of fractionated protein sample per ZOOM Strip. Use an accurate and sensitive protein estimation method.
*Insensitive detection method: Use sensitive detection methods such as silver staining or immunoblotting.

I used the ZOOM IPGRunner system and got vertical streaking. Can you please offer some suggestions?

Here are the possible causes and solutions:

*Protein has got oxidized: Include DTT in the rehydration buffer and perform the alkylation step.
*Impure solutions: Use ultrapure reagents to prepare the rehydration buffer, equilibration buffer, and buffers for SDS/PAGE.

I used the ZOOM IPGRunner System and got horizontal streaking. Can you please offer some suggestions?

Here are the possible causes and solutions:

*Impure solutions: Use ultrapure reagents to prepare the rehydration buffer, equilibration buffer, and buffers for SDS/PAGE.
*Air bubble between the strip and 2D gel: Smooth out the air bubbles.
*Poor strip rehydration: Rehydrate the strips in 140 µL rehydrating buffer for 1 hour as described in the manual. Rehydration can be extended to overnight if you use 155 µL rehydrating buffer. Make sure that the rehydration buffer covers the strip completely.
*Protein overload: Decrease the protein concentration or lower the sample volume.
*Protein precipitates: Increase solubilization reagents in the rehydration buffer (see manual). Use appropriate strips based on the pI of the protein sample. Do not add more than 10 µL of your sample to 140 µL of rehydration buffer (see manual).

I used the ZOOM IPGRunner System and got incomplete focusing of my proteins. Can you please offer some tips?

Here are the possible causes and solutions:

*Improper sample preparation: Increase solubilization reagents in the rehydration buffer. Use 8 M urea for solubilization. Add DTT and non-ionic detergents as described in the manual.
*Incorrect focusing time: Increase or decrease the focusing time based on the initial results.
*High protein load: Decrease the protein load. Use an accurate and sensitive protein estimation method.
*Salt, lipids, or nucleic acid impurities present in the sample: Remove interfering substances (see manual for details).
*Poor strip rehydration: Rehydrate the strips in 140 µL rehydrating buffer for 1 hour as described in the manual. Rehydration can be extended to overnight if you use 155 µL rehydrating buffer. Make sure the rehydration buffer is covering the strip completely.

After the first dimension separation using the ZOOM IPGRunner System, I do not see any distinct spots. What could the issue be?

Here are the possible causes and solutions:

*Low protein load. Increase the protein load. Use an accurate and sensitive protein estimation method.
*Improper sample preparation. Increase solubilization reagents. Use at least 8 M urea for solubilization. Add DTT and non-ionic detergents (see manual for details).
*Strip not correctly oriented. Align the strip correctly as described in the manual. Be sure to have the gel side up when loading the strip into the ZOOM IPGRunner Cassette.
*Air bubbles between the strip and 2D gel. Smooth out any air bubbles.
*Insensitive detection method. Use sensitive detection methods such as silver staining or immunoblotting.

What are the power supply considerations for the ZOOM IPGRunner system?

The ZOOM IPGRunner System should be used with an external DC power supply designed for IEF and electrophoresis applications. This power supply must:

*Be isolated from the ground so that the DC output is floating.
*Be programmable, with a 4-protocol minimum.
*Be able to operate at low current (<1 mA) as IEF is performed at very low current.
Note: Many power supplies automatically shut off when the current drops below 1 mA. You will need a power supply capable of overriding the low current shut-off feature.

The electrical leads of the ZOOM IPGRunner Lid are recessed and may not fit into some power supply units. To allow connection of the ZOOM IPGRunner power leads with certain power supplies, use Invitrogen Power Supply Adapters available separately (Cat. No. ZA10001).

What is the optimal concentration of detergent that I should add to my sample for the ZOOM IPG Runner?

Detergents are used in the sample as solubilizing agents:

*SDS must be 0.2% or lower. If the percentage of SDS is too high, all the proteins will be negatively charged and will migrate to the positive electrode.
*CHAPS can be increased up to 4%.
*NP-40, Triton-X and other nonionic detergents are used at 0.5-0.7%.

What is the optimal salt concentration that I should use in my sample for the ZOOM IPG Runner?

The salt concentration should not exceed 10 mM for these reasons:

*Burning can occur when conductivity is high, hence current limits should be set.
*Protein transport in an electric field decreases with increasing ionic strength.
*The efficiency of all IEF instruments is decreased when samples contain more than 10 mM salt.
*Longer run times are generally required for samples containing >10 mM salt.

Can I run non-reduced samples in the ZOOM IPGRunner?

You can run non-reduced samples in the ZOOM IPGRunner by omitting DTT in the rehydration buffer and equilibration buffers.

Can I heat my protein sample prepared using ZOOM 2D Protein Solubilizer?

We do not recommend heating protein samples containing urea over 37 degrees C as elevated temperatures cause urea to hydrolyze to isocyanate, which modifies proteins by carbamylation.

How do you recommend storing protein samples prepared in the Sample Rehydration buffer?

We recommend storing them at -80 degrees C. We do not recommend storing them at -20 degrees C

How much ampholyte do you recommend adding to the sample rehydration buffer?

The recommended ampholyte concentration in the sample rehydration buffer is 0.5%.

*If you are loading 5-50 µg of protein (pure protein or crude lysate) per ZOOM strip, use 0.5% ampholytes in the sample rehydration buffer.
*If you are loading >50 µg of protein (crude lysate or fractionated sample) per ZOOM Strip, use 0.5-2% ampholytes in the sample rehydration buffer.

Note: Higher ampholyte concentration requires longer focusing times.

What is the maximum recommended volume of protein sample to be added to the Sample Rehydration buffer?

The maximum volume of the protein sample should at most be 1/6 of the final sample volume that will be added to the strip. A good general volume would be 5-10µL. 140 µL of sample diluted in Sample Rehydration buffer is used to rehydrate each ZOOM Strip for the standard rehydration time of one hour. For overnight rehydration, we recommend using 155 µL of diluted sample.

I am using your ZOOM IPGRunner system. What is the sample rehydration buffer?

The sample rehydration buffer, also known as the sample buffer, is used to denature and solubilize protein samples, and rehydrate the IPG strips prior to performing IEF using the ZOOM IPGRunner system. Due to the large variety of proteins, there is no universal sample rehydration buffer. The sample rehydration buffer must maintain proteins in solution during rehydration of the IPG strips and IEF, and must not have any effect on the pI of the protein. The buffer typically contains a denaturing agent (urea or urea/thiourea), solubilizing agent (non-ionic or zwitterionic detergent and ampholytes), and reducing agent (DTT).

Note: To obtain high resolution, IEF is usually performed under denaturing and reducing conditions. Use a denaturing sample rehydration buffer for sample preparation (see manual for recipe).

How does the ZOOM IPG Runner System work?

The ZOOM IPGRunner System provides a convenient and quick way to perform isoelectric focusing (IEF) of proteins in a vertical mini-gel format using immobilized pH gradient (IPG) strips for two- dimensional (2D) gel electrophoresis.

The major components of the ZOOM IPGRunner System are:

*ZOOM IPGRunner Mini-Cell (Cat. No. ZM0001)
*ZOOM IPGRunner Cassettes (1 pack of 10 cassettes, Cat. No. ZM0003)
*ZOOM Strips (12 ZOOM Strips, pH 3–10 NL, Cat. No. ZM0011)

Using the ZOOM Strips, proteins are separated based on their isoelectric point (pI). The proteins in the sample can be further separated in the second dimension, based on their molecular weight, by placing the IPG Strips into the IPG well of a NuPAGE Bis-Tris ZOOM gel or Tris-Glycine ZOOM gel. The proteins separated in the second dimension can be visualized as spots by the use of SimplyBlue SafeStain or silver staining, or blotted onto membranes. Protein spots can be also excised from the gel or the membranes to be further analyzed by mass spectrometry or chemical microsequencing to facilitate protein identification.

What is the purpose of alkylation prior to IEF?

Alkylation prevents unwanted protein modifications by alkylating cysteines to avoid mixed disulfide formation and reoxidation and this allows for crisper focusing.

Do you still offer ZOOM 2D Protein Solubilizers?

Sorry we have discontinued selling the ZOOM 2D Protein Solubilizers.

What are the main reasons for performing 2D gel electrophoresis of proteins?

The main advantages of performing 2D gel electrophoresis of proteins and applications used for are listed below:

Advantages:

*Simultaneous separation of hundreds to thousands of proteins
*High capacity with superior resolution
*Compatible with further analysis by MS for protein identification and sequencing
Ability to separate and analyze low-abundance proteins

Applications:

*Comparative proteomics: identifying and analyzing differences between complex mixtures of proteins
*Protein profiling, biomarker discovery
*Separation and analysis of protein variants and isoforms

What products do you offer for 2D gel electrophoresis of proteins?

We offer the following products for the first- and second-dimension separation of proteins:

First-dimension separation:

*Vertical gels for separation of proteins based on their isoelectric point (pI) (https://www.thermofisher.com/us/en/home/life-science/protein-expression-and-analysis/protein-gel-electrophoresis/protein-gels/specialized-protein-separation/isoelectric-focusing.html)

*Solution-phase isoelectric focusing of proteins using ZOOM Disks (immobilized buffer disks of specific pH) to reduce sample complexity, enrich low-abundance proteins, and increase the dynamic range of detection (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/isoelectric-focusing/zoom-ief-fractionator.html)

*Mini gel system for high-throughput isoelectric focusing of proteins using ZOOM IPG (Immobilized pH Gradient) Strips (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/2d-gel-electrophoresis/zoom-ipgrunner-system.html)

Second-dimension separation:

*ZOOM gels for 2D electrophoresis: NuPAGE Bis-Tris (Cat. No. NP0330BOX) and Tris-Glycine (Cat. No. EC60261BOX) mini gels with IPG wells ( to accommodate 7 cm ZOOM strips) for separation of proteins based on their molecular weight

What is 2D protein gel electrophoresis?

2D protein gel electrophoresis is the separation of proteins in two dimensions. In the first dimension, proteins are separated by their isoelectric point (pI) using isoelectric focusing, and in the second dimension, they are separated by their mass using SDS-PAGE.


For Research Use Only. Not for use in diagnostic procedures.