missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ IPGRunner™ Cassette DFS Item

For use with the ZOOM IPGRunner Mini-Cell

Supplier:  Invitrogen™ ZM0003

Catalog No. ZM0003


Only null left
Explore available promotions

Includes: Electrode wicks and sealing tape.

Description

Description

The ZOOM IPGRunner Cassette provides a convenient way to rehydrate 7.0cm ZOOM Strips and perform isoelectric focusing of up to six 7.0cm ZOOM Strips (IPG strips) without requiring separate units for rehydration and isoelectric focusing.

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

• Zoom IPGRunner Cassettes (10)
• Sealing tape (22)
• Electrode wicks (22)

IPGRunner, ZOOM
Room Temperature
ZOOM IPGRunner System
10 cassette(s)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.