missing translation for 'onlineSavingsMsg'
Learn More

Zoledronic Acid Monohydrate 98.0+%, TCI America™

Catalog No. Z00315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z00315G 5 g
Z00311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z00315G Supplier TCI America Supplier No. Z00315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 165800-06-6
Molecular Formula C5H12N2O8P2
Molecular Weight (g/mol) 290.105
MDL Number MFCD00867791
InChI Key FUXFIVRTGHOMSO-UHFFFAOYSA-N
Synonym 1-Hydroxy-2-(1-imidazolyl)ethane-1,1-diphosphonic Acid
PubChem CID 121586
IUPAC Name (1-hydroxy-2-imidazol-1-yl-1-phosphonoethyl)phosphonic acid;hydrate
SMILES C1=CN(C=N1)CC(O)(P(=O)(O)O)P(=O)(O)O.O

Specifications

Melting Point 245°C
Color White-Yellow
Quantity 5 g
Formula Weight 272.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Zoledronic Acid Monohydrate
RTECSNumber SZ8562450
TSCA No
Recommended Storage Refrigerator