missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ZNF521 Antibody Blocking Peptide

Catalog No. NB398496
Change view
Click to view available options
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NB398496 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398496 Supplier R&D Systems™ Supplier No. NBP191271PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Blocking Peptide

Specifications

Gene ID (Entrez) 25925
Product Line Novus
Concentration 1 mg/ml
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Use With (Application) This peptide is useful as a blocking peptide for NBP1-91271. For further blocking peptide related protocol, click here .
Gene Alias EHZF, Evi3, LIP3, ZFP521, zinc finger protein 263, Zinc finger protein FPM315, Zinc finger protein with KRAB and SCAN domains 12
Gene Symbol ZNF521
Quantity 0.1 mg
Research Category Immunology, Neuroscience
Storage Buffer Peptide dissolved in dH20. Contains no BSA.