missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Fluoride, Powder, USP, EP, BP, 98.5-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Fluoride, Powder, USP, EP, BP is used to prevent cavities making teeth stronger and more resistant to decay caused by acid and bacteria. Toothpaste being the preferred delivery method. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Powder, Needles or Crystals |
| Quantity | 125 g |
| Percent Purity | 98.5 to 100.5% |
| Grade | BP/EP/USP |
| Loss on Drying | 0.005 |
| Packaging | Poly Bottle |
| Appearance | Clear |
| Chemical Name or Material | Sodium Fluoride, Powder |