MW 656.68 g/mol, Purity >90%. Cell permeable derivative of ZnAF-2. Fluorescent reagent employed in the detection of zinc ion (after esterase hydrolysis).
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 656.68 g/mol, Purity >90%. Cell permeable derivative of ZnAF-2. Fluorescent reagent employed in the detection of zinc ion (after esterase hydrolysis).