Learn More
Abcam Inc ZnAF-2 DA, Fluorescent Zn2+ probe, 5MG
Catalog No. 502705203
Encompass
MW 656.68 g/mol, Purity >90%. Cell permeable derivative of ZnAF-2. Fluorescent reagent employed in the detection of zinc ion (after esterase hydrolysis).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705203 Supplier Abcam Inc Supplier No. AB1454225MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 656.68 g/mol, Purity >90%. Cell permeable derivative of ZnAF-2. Fluorescent reagent employed in the detection of zinc ion (after esterase hydrolysis).
The product is subject to the following: Abcam Restricted Use Statement