missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconocene Bis(trifluoromethanesulfonate) Tetrahydrofuran Adduct 97.0+%, TCI America™
$259.11 - $1,002.86
Chemical Identifiers
| CAS | 89672-77-5 |
|---|---|
| Molecular Formula | C16H18F6O7S2Zr |
| MDL Number | MFCD00192583 |
| Synonym | Bis(cyclopentadienyl)zirconium Bis(trifluoromethanesulfonate), Zirconocene Bis(triflate) |
Chemical Identifiers
| 89672-77-5 | |
| C16H18F6O7S2Zr | |
| MFCD00192583 | |
| Bis(cyclopentadienyl)zirconium Bis(trifluoromethanesulfonate), Zirconocene Bis(triflate) |
Specifications
| 89672-77-5 | |
| MFCD00192583 | |
| 1325 | |
| 591.65 | |
| Crystalline Powder |
| C16H18F6O7S2Zr | |
| 1 g | |
| Bis(cyclopentadienyl)zirconium Bis(trifluoromethanesulfonate), Zirconocene Bis(triflate) | |
| ≥97.0% (T) | |
| Zirconocene Bis(trifluoromethanesulfonate) Tetrahydrofuran Adduct |
Safety and Handling
TSCA : No