missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconocene Bis(trifluoromethanesulfonate) Tetrahydrofuran Adduct 97.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| UN Number | 1325 |
| Formula Weight | 591.65 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Zirconocene Bis(trifluoromethanesulfonate) Tetrahydrofuran Adduct |
Safety and Handling
| TSCA | No |