missing translation for 'onlineSavingsMsg'
Learn More

Zirconium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis excluding Hf), Hf nominal 3%, Thermo Scientific Chemicals

Catalog No. AA42556QK
Change view
Click to view available options
Quantity:
25 Slugs
5 Slugs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42556QK 25 Slugs
AA42556QJ 5 Slugs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42556QK Supplier Thermo Scientific Chemicals Supplier No. 042556QK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: Approx.2.62g/slug

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Color Gray to Silver
Appearance Gray to silver slug
Quantity 25 Slugs
Odor Odorless
Form Slug
Assay Percent Range 99.95% (Metals basis excluding Hf)
Chemical Name or Material Zirconium
EINECSNumber 231-176-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only