missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.95% (metals basis excluding Hf), Hf nominal 3%, Thermo Scientific Chemicals
$601.97 - $601.97
Chemical Identifiers
| CAS | 7440-67-7 |
|---|---|
| Molecular Formula | Zr |
| Molecular Weight (g/mol) | 91.22 |
| MDL Number | MFCD00011303 MFCD00148878 |
| InChI Key | QCWXUUIWCKQGHC-UHFFFAOYSA-N |
| Synonym | zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil |
| PubChem CID | 23995 |
| ChEBI | CHEBI:33342 |
| IUPAC Name | zirconium |
| SMILES | [Zr] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-67-7 | |
| 91.22 | |
| QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
| 23995 | |
| zirconium |
| Zr | |
| MFCD00011303 MFCD00148878 | |
| zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
| CHEBI:33342 | |
| [Zr] |
Specifications
| 7440-67-7 | |
| Odorless | |
| Slug | |
| MFCD00011303 MFCD00148878 | |
| QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
| zirconium | |
| 23995 | |
| 99.95% | |
| Zirconium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length |
| 25 Slugs | |
| ≈1.31g/slug | |
| Zr | |
| zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
| [Zr] | |
| 91.22 | |
| CHEBI:33342 | |
| Hf nominal 3% | |
| (metals basis excluding Hf), Hf nominal 3% |
Safety and Handling
EINECSNumber : 231-176-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only