missing translation for 'onlineSavingsMsg'
Learn More

Zirconium plate, 6.4mm (0.25in) thick, 25x100mm (0.98x3.9in), 99.2% (metals basis excluding Hf), Thermo Scientific Chemicals

Catalog No. AA00934KF
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA00934KF 1 Pc.
AA00934KJ 5 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA00934KF Supplier Thermo Scientific Chemicals Supplier No. 000934KF

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Quantity 1 Pc.
Odor Odorless
Weight ≈104g/25x100mm
Form Plate
Percent Purity 99.2%
Assay (metals basis excluding Hf)
Chemical Name or Material Zirconium plate, 6.4mm (0.25 in.) thick, 25x100mm (0.98x3.9 in.), Temper: annealed
EINECSNumber 231-176-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only