missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide Yttrium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals

$176.53 - $734.37

Specifications

Chemical Name or Material Zirconium Oxide Yttrium Oxide Substrate
Name Note Polished One Side
Physical Form Plate
Molecular Formula ZrO2 + 9.5% Y2O3
MDL Number MFCD01868885
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38509KT
View Documents
Thermo Scientific Chemicals
038509KT
1 Ea. N/A
Only null left
 
AA38509KU
View Documents
Thermo Scientific Chemicals
038509KU
5 Ea. N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zirconium Oxide Yttrium Oxide Substrate
Plate
MFCD01868885
Polished One Side
ZrO2 + 9.5% Y2O3
Odorless
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only