missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide, catalyst support, Thermo Scientific Chemicals

$175.48 - $2,393.75

Chemical Identifiers

MDL Number MFCD00011310
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4381422
View Documents
Thermo Scientific Chemicals
04381422
100 g N/A
Only null left
 
AA4381436
View Documents
Thermo Scientific Chemicals
04381436
500 g N/A
Only null left
 
AA43814A4
View Documents
Thermo Scientific Chemicals
043814A4
2.5 kg N/A
Only null left
 
Description

Description

Catalyst carrier

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00011310

Specifications

Pellets
100 g
MFCD00011310
Zirconium Oxide, Catalyst Support
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-227-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only