missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide, catalyst support, monoclinic phase, Thermo Scientific Chemicals

Catalog No. AA4381522
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
2.5 kg
500 g
Catalog No. Quantity
AA4381522 100 g
AA4381536 500 g
AA43815A4 2.5 kg
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Catalog No. AA4381522

Supplier: Thermo Scientific Chemicals 04381522

Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Typical properties S.A. 90m2/g, median pore diameter 80/400Å, bimodal total pore volume 0.31cc/g, packing density 1.181g/L

Zirconium oxide is used as a catalyst carrier, diamond simulant, refractory material, pigment, abrasives and solid electrolyte in electrochromic devices. It acts as a precursor to the electroceramic lead zirconate titanate. It is also used in insulation, oxygen sensors, enamels and fuel cell. It finds application to make ceramic knives due to its hardness nature. It is utilized for dental restorations like crowns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00011310

Specifications

Physical Form Pellets
Quantity 100 g
Solubility Information Soluble in hot-concentrated hydrogen fluoride and sulfuric acid. Insoluble in water.
Chemical Name or Material Zirconium oxide, catalyst support, 1/8 in. Pellets
Hazard Category H500
EINECSNumber 215-227-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only