missing translation for 'onlineSavingsMsg'
Learn More

Zirconium n-butoxide, 80% w/w in 1-butanol, Thermo Scientific Chemicals

Catalog No. AAB2331718
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2331718 50 g
AAB2331730 250 g
AAB233170B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2331718 Supplier Thermo Scientific Chemicals Supplier No. B2331718
Only null left

Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, acids, bases, halogens, alkali metals, water/ moisture.

Chemical Identifiers

CAS 1071-76-7
Molecular Formula C16H40O4Zr+4
Molecular Weight (g/mol) 387.716
MDL Number MFCD00048765
InChI Key ASUYWGMDIAPAKI-UHFFFAOYSA-N
Synonym 1-butanol, zirconium 4+ salt 4:1, 1-butanol, zirconium 4++ salt
PubChem CID 122130442
IUPAC Name butan-1-ol;zirconium(4+)
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Zr+4]

Specifications

Quantity 50 g
Linear Formula [CH3(CH2)3O]4Zr
UN Number UN1120
Beilstein 4132649
Flash Point 38°C (100°F)
Solubility Information Hydrolyzes in water.
Formula Weight 383.68
Concentration or Composition (by Analyte or Components) 80% w/w in 1-butanol
Odor Alcohol-like
Refractive Index 1.466
Sensitivity Moisture sensitive
Density 1.07
Chemical Name or Material Zirconium n-butoxide
Show More Show Less
Hazard Category H226-H315-H318-H335-H336
Hazard Statement GHS H Statement
H318-H226-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapour.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
EINECSNumber 213-995-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only