missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7058760
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4319414 25 g
AA4319422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4319414 Supplier Thermo Scientific Chemicals Supplier No. 04319414
Only null left

Used in catalysts, precipitation of amino acids and proteins, and tanning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7446-31-3
Molecular Formula H8O12S2Zr
Molecular Weight (g/mol) 355.40
MDL Number MFCD00149267
InChI Key NLOQZPHAWQDLQW-UHFFFAOYSA-J
Synonym zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfate
PubChem CID 23219663
IUPAC Name zirconium(4+);disulfate;tetrahydrate
SMILES O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Merck Index 14,10182
Solubility Information Soluble in water (525g/L).
Formula Weight 355.40 (283.34 Anhydrous)
Percent Purity 99.99%
Density 3.22 g/mL
Assay (metals basis)
Chemical Name or Material Zirconium (IV) sulfate tetrahydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-694-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only