Learn More
Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals
Catalyst support
Supplier: Thermo Scientific Chemicals 04104136
Description
Catalyst support.Zirconium disulfate tetrahydrate is used in organic synthesis. It is also used in tanning white leather. Further, it is used as catalyst supports to precipitate proteins and amino acids. It is also useful in chemical analysis. It plays an essential role as an additive for lubricants and as a pigment stabilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Zirconium(IV) sulfate tetrahydrate | |
| 7446-31-3 | |
| (metals basis) | |
| MFCD00149267 | |
| zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfate | |
| NLOQZPHAWQDLQW-UHFFFAOYSA-J | |
| zirconium(4+);disulfate;tetrahydrate | |
| 23219663 | |
| ≥98% |
| White | |
| 500 g | |
| H8O12S2Zr | |
| 14,10182 | |
| Soluble in water. | |
| O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 355.40 | |
| 355.40 (283.34 Anhydrous) | |
| 3.22 g/mL |
Chemical Identifiers
| 7446-31-3 | |
| 355.40 | |
| NLOQZPHAWQDLQW-UHFFFAOYSA-J | |
| 23219663 | |
| O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| H8O12S2Zr | |
| MFCD00149267 | |
| zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfate | |
| zirconium(4+);disulfate;tetrahydrate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-694-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only