missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, nanopowder, 99% (metals basis excluding Hf), Thermo Scientific Chemicals

Catalog No. p-7059549
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4488614 25 g
AA4488622 100 g
AA4488636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4488614 Supplier Thermo Scientific Chemicals Supplier No. 04488614
Only null left

Zirconium(IV) oxide is used as an additive in the production of piezoelectric crystals, in high-frequency induction coils and in ceramic glazes. It finds application as solid electrolyte in electrochromic devices and precursor to electroceramic lead zirconate titanate (PZT), as insulator in transistors, as a thermal barrier coating in jet and diesel engines and as diamond simulant in jewelry. It is used in oxygen sensors, fuel cell membranes and in orthopaedic implants of hip joints. It is coated on silica gel surface and found to be used in the extraction of chromium from aqueous solutions at a pH of about 3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Boiling Point ∼5000°C
Melting Point ∼2700°C
Color White
Physical Form Powder
Quantity 25 g
Linear Formula ZrO2
Merck Index 14,10180
Solubility Information Insoluble in water. Soluble in sulfuric acid and hydrofluoric acid.
Formula Weight 123.22
Percent Purity 99%
Odor Odorless
Density 5.89 g/mL
Assay (metals basis excluding Hf)
Chemical Name or Material Zirconium (IV) oxide
Show More Show Less
Hazard Category H500
EINECSNumber 215-227-2
RTECSNumber ZH8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only