missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 99.7% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Catalog No. p-4904584
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3631918 50 g
AA3631930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3631918 Supplier Thermo Scientific Chemicals Supplier No. 03631918
Only null left

For vacuum deposition

For vacuum deposition. Raw material for the synthesis. Used as Additive of special glass, ceramics. It is also used as a detector of alpha-rays, which render it luminescent. As vacuum coating. Piezoelectric ceramics include filter ,speaker, the ultrasonic underwater acoustic detectors and so on . Stabilized oxide For refractory furnace linings, and in crucibles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Boiling Point ca 5000°C
Melting Point ∽2700°C
Color White
Physical Form Sintered Lump
Quantity 50 g
Assay Percent Range 99.7% metal basis excluding Hf
Solubility Information Insoluble in water. Soluble in hot and concentrated hydrofluoric acid and sulfuric acid. Dissolves on heating in a mixture of 2 parts H2SO4 and 1 part water.
Formula Weight 123.22
Odor Odorless
Density 5.89
Chemical Name or Material Zirconium(IV) oxide, Hf < 75ppm
Show More Show Less
EINECSNumber 215-227-2
RTECSNumber ZH8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only