missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 99.7% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04152830

 View more versions of this product

Catalog No. AA4152830


Only null left
Explore available promotions

Description

Description

Zirconium(IV) oxide is used as an additive in the production of piezoelectric crystals, in high-frequency induction coils and in ceramic glazes. It finds application as solid electrolyte in electrochromic devices and precursor to the electroceramic lead zirconate titanate (PZT), as insulator in transistors, as a thermal barrier coating in jet and diesel engines and as diamond simulant in jewelry. It is used in oxygen sensors, fuel cell membranes and in orthopaedic implants of hip joints. It is coated on silica gel surface and found to be used in the extraction of chromium from aqueous solutions at a pH of about 3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

∼5000°C
∼2700°C
Powder
(metals basis excluding Hf), Hf <75ppm
MFCD00011310 MFCD01868884
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
99.7%
5.89 g/mL
Zirconium(IV) oxide
1314-23-4
250 g
O2Zr
14,10180
Soluble in alcohols,hydrofluoric and hot sulphuric acid. Insoluble in water.
O=[Zr]=O
123.22
123.22
Odorless

Chemical Identifiers

1314-23-4
123.22
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00011310 MFCD01868884
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
dioxozirconium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-227-2

RTECSNumber : ZH8800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only